Learn More
LGC GENOMICS LLC (Lucigen) CopyCutter EPI400 Electrocompetent E. coli, 10 RxnsClone unstable and toxic sequences in standard plasmids, with high efficiency.
Supplier:  LGC GENOMICS LLC C400EL10
Encompass
CopyCutter™ EPI400™ E. coli cells will significantly lower the copy number of a wide variety of common vectors to enable cloning of unstable DNA sequences and those expressing toxic proteins
Catalog No. NC1242118