Learn More
ABclonal Technology COQ10B Rabbit pAb
SDP
Catalog No. 5000108668
Encompass_Preferred
Predicted to enable ubiquinone binding activity. Predicted to be involved in cellular respiration and ubiquinone biosynthetic process. Predicted to be located in mitochondrial inner membrane. Predicted to be active in mitochondrion.
Catalog No. 50-001-08668 Supplier ABclonal Technology Supplier No. A15193500UL
Predicted to enable ubiquinone binding activity. Predicted to be involved in cellular respiration and ubiquinone biosynthetic process. Predicted to be located in mitochondrial inner membrane. Predicted to be active in mitochondrion.