Learn More
Aviva Systems Biology Corp COQ7 Antibody - C-terminal region (ARP64009_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COQ7. It was validated on Western Blot. . Alternate names: CAT5, CLK-1, CLK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-288-139