Learn More
Aviva Systems Biology Corp COQ9 Antibody - N-terminal region (ARP63366_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63366_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COQ9. It was validated on Western Blot. . Alternate names: C16orf49, DKFZp434K046ApplicationWB, IHC. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-949