Learn More
ABclonal Technology COQ9 Rabbit pAb
SDP
Catalog No. 5000104524
Encompass_Preferred
This locus represents a mitochondrial ubiquinone biosynthesis gene. The encoded protein is likely necessary for biosynthesis of coenzyme Q10, as mutations at this locus have been associated with autosomal-recessive neonatal-onset primary coenzyme Q10 deficiency.
Catalog No. 50-001-04524 Supplier ABclonal Technology Supplier No. A15480500UL
This locus represents a mitochondrial ubiquinone biosynthesis gene. The encoded protein is likely necessary for biosynthesis of coenzyme Q10, as mutations at this locus have been associated with autosomal-recessive neonatal-onset primary coenzyme Q10 deficiency.