Learn More
ABclonal Technology COQ9 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154801000UL
Encompass_Preferred
This locus represents a mitochondrial ubiquinone biosynthesis gene. The encoded protein is likely necessary for biosynthesis of coenzyme Q10, as mutations at this locus have been associated with autosomal-recessive neonatal-onset primary coenzyme Q10 deficiency.
Catalog No. 50-001-05626