Learn More
ACCEGEN BIOTECH COR-L279 cell line, ≥1.0 million/ frozen vial
Catalog No. NC3659655
Encompass
Human lung small cell carcinoma. COR-L279 has been isolated from a lymph node of an untreated 63 year-old male. The degenerated and necrotic morphology of the sample obtained was consistent with metastatic small cell carcinoma of the lung.
Catalog No. NC3659655 Supplier ACCEGEN BIOTECH Supplier No. ABCTC0181
Human lung small cell carcinoma. COR-L279 has been isolated from a lymph node of an untreated 63 year-old male. The degenerated and necrotic morphology of the sample obtained was consistent with metastatic small cell carcinoma of the lung.