Learn More
Grainger Cor Rad End Mill 1/16 Carb 0.0100 rad
Catalog No. 502749738
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch overall length. The end mill has a TiAlN coating and 4 flutes. It is designed for high speed machining in aluminum, brass, bronze, copper, plastics, and other non-ferrous materials. The end mill provides a strong, sharp cutting edge and smooth corner finish. With its small corner radius, it is well-suited for machining tight internal radii.
Catalog No. 50-274-9738 Supplier Grainger Supplier No. 14R973
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch overall length. The end mill has a TiAlN coating and 4 flutes. It is designed for high speed machining in aluminum, brass, bronze, copper, plastics, and other non-ferrous materials. The end mill provides a strong, sharp cutting edge and smooth corner finish. With its small corner radius, it is well-suited for machining tight internal radii.