Learn More
Grainger Cor Rad End Mill 1/16 Carb 0.0050 rad
Catalog No. 502749741
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has a TiAlN coating and 4 flutes. It is designed for high feed milling in steel, stainless steel, and cast iron. The end mill provides good chip evacuation and can mill small radii.
Catalog No. 50-274-9741 Supplier Grainger Supplier No. 14R962
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has a TiAlN coating and 4 flutes. It is designed for high feed milling in steel, stainless steel, and cast iron. The end mill provides good chip evacuation and can mill small radii.