Learn More
Grainger Cor Rad End Mill 1/16 Carb 0.0100 rad
Catalog No. 502749853
Encompass_Preferred
The Corner Radius End Mill by Niagara Cutter is a high-performance end mill designed for milling corners and edges with a radius. It features a corner radius of 0.031 inches and an overall length of 2.5 inches. The end mill has 4 flutes and is made of solid carbide for durability and long tool life. It has a neck relief to provide strength in high load applications. The end mill has a TiAIN coating to resist heat and built-up edge. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and nickel alloys. The end mill is center cutting and provides excellent chip control when milling corners and edges.
Catalog No. 50-274-9853 Supplier Grainger Supplier No. 14R969
The Corner Radius End Mill by Niagara Cutter is a high-performance end mill designed for milling corners and edges with a radius. It features a corner radius of 0.031 inches and an overall length of 2.5 inches. The end mill has 4 flutes and is made of solid carbide for durability and long tool life. It has a neck relief to provide strength in high load applications. The end mill has a TiAIN coating to resist heat and built-up edge. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and nickel alloys. The end mill is center cutting and provides excellent chip control when milling corners and edges.