Learn More
Grainger Cor Rad End Mill 1/16 Carb 0.0100 rad
Catalog No. 502752127
Encompass_Preferred
The Micro 100 Corner Radius End Mill Carbide 14R972 is a high-performance end mill designed for machining corners and edges. It features a 0.125" corner radius and a 0.500" cutting diameter. The end mill has 4 flutes and is made of solid carbide for durability and long tool life. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and high-temp alloys. The 14R972 has AlTiN nano coating to resist heat and built-up edge. It is well-suited for 2D and 3D profiling, slotting, side milling, and contouring applications requiring sharp corners and edges.
Catalog No. 50-275-2127 Supplier Grainger Supplier No. 14R972
The Micro 100 Corner Radius End Mill Carbide 14R972 is a high-performance end mill designed for machining corners and edges. It features a 0.125" corner radius and a 0.500" cutting diameter. The end mill has 4 flutes and is made of solid carbide for durability and long tool life. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and high-temp alloys. The 14R972 has AlTiN nano coating to resist heat and built-up edge. It is well-suited for 2D and 3D profiling, slotting, side milling, and contouring applications requiring sharp corners and edges.