Learn More
Grainger Cor Rad End Mill 1/16 Carb 0.0050 rad
Catalog No. 502748851
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.002 to 0.012 IPR. It is well-suited for machining corners and edges in steels, stainless steels, cast iron, aluminum and other non-ferrous metals. The end mill features Micro 100's proprietary AM100 coating for increased tool life and performance.
Catalog No. 50-274-8851 Supplier Grainger Supplier No. 14R967
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.002 to 0.012 IPR. It is well-suited for machining corners and edges in steels, stainless steels, cast iron, aluminum and other non-ferrous metals. The end mill features Micro 100's proprietary AM100 coating for increased tool life and performance.