Learn More
Grainger Cor Rad End Mill 1/2 Carb 0.0100 rad
Catalog No. 502751878
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish end mill made of solid carbide with a corner radius of 0.031". It has 2 flutes and a right hand spiral. The overall length is 2.5" and shank diameter is 0.25". It is designed for high feed milling of aluminum, brass, bronze, copper, plastic and other non-ferrous metals. This end mill provides a strong, precision cutting edge and smooth milling action. Its bright finish resists corrosion and reduces heat buildup.
Catalog No. 50-275-1878 Supplier Grainger Supplier No. 14T093
The Micro 100 Corner Radius End Mill is a bright finish end mill made of solid carbide with a corner radius of 0.031". It has 2 flutes and a right hand spiral. The overall length is 2.5" and shank diameter is 0.25". It is designed for high feed milling of aluminum, brass, bronze, copper, plastic and other non-ferrous metals. This end mill provides a strong, precision cutting edge and smooth milling action. Its bright finish resists corrosion and reduces heat buildup.