Learn More
Grainger Cor Rad End Mill 1/2 Carb 0.0200 rad
Catalog No. 502752933
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a 14T089 model milling cutter with a square end for machining flat-bottomed cavities and slots. It has a 1/2 inch shank diameter and is made of solid carbide for wear resistance and strength. The overall length is 3 inches and it has 2 straight flutes. The mill has an uncoated carbide grade of C-2 for machining aluminum and other non-ferrous materials. It can be run at speeds up to 2000 SFM. This is a high quality carbide end mill for light milling applications.
Catalog No. 50-275-2933 Supplier Grainger Supplier No. 14T089
The Micro 100 Square End Mill Carbide is a 14T089 model milling cutter with a square end for machining flat-bottomed cavities and slots. It has a 1/2 inch shank diameter and is made of solid carbide for wear resistance and strength. The overall length is 3 inches and it has 2 straight flutes. The mill has an uncoated carbide grade of C-2 for machining aluminum and other non-ferrous materials. It can be run at speeds up to 2000 SFM. This is a high quality carbide end mill for light milling applications.