Learn More
Grainger Cor Rad End Mill 1/32 Carb 0.0050 rad
Catalog No. 502748866
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a 2 flute design. The overall length is 2 inches and it has a 0.125 inch max depth of cut. This end mill is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. It features a TiAlN coating for increased tool life and heat resistance. The corner radius profile allows for strong cutting edges and smooth wall blends.
Catalog No. 50-274-8866 Supplier Grainger Supplier No. 14R959
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a 2 flute design. The overall length is 2 inches and it has a 0.125 inch max depth of cut. This end mill is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. It features a TiAlN coating for increased tool life and heat resistance. The corner radius profile allows for strong cutting edges and smooth wall blends.