Learn More
Grainger Cor Rad End Mill 1/32 Carb 0.0050 rad
Catalog No. 502752134
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM. It is made of solid carbide for wear resistance when milling steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. This end mill is designed for high performance slotting, contouring, and profiling applications.
Catalog No. 50-275-2134 Supplier Grainger Supplier No. 14R961
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM. It is made of solid carbide for wear resistance when milling steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. This end mill is designed for high performance slotting, contouring, and profiling applications.