Learn More
Grainger Cor Rad End Mill 1/32 Carb 0.0050 rad
Catalog No. 502749646
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is made of solid carbide for wear resistance and strength. The end mill has a TiAIN coating for heat resistance and lubricity. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum and other non-ferrous materials. The end mill has 2 flutes and a right hand spiral. It is manufactured in the USA by Micro 100 Tool Corporation.
Catalog No. 50-274-9646 Supplier Grainger Supplier No. 14R958
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is made of solid carbide for wear resistance and strength. The end mill has a TiAIN coating for heat resistance and lubricity. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum and other non-ferrous materials. The end mill has 2 flutes and a right hand spiral. It is manufactured in the USA by Micro 100 Tool Corporation.