Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0100 rad
Catalog No. 502751102
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a high performance end mill designed for machining corners and edges. It features a corner radius of 0.031" and is made of solid carbide with TiAlN coating for wear resistance and heat dissipation. This 1/4" shank end mill has 2 flutes and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and high temp alloys. It has a center cutting design allowing plunge cuts and can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. The end mill has a reach of 1" and overall length of 2-1/2".
Catalog No. 50-275-1102 Supplier Grainger Supplier No. 14T029
The Micro 100 Corner Radius End Mill is a high performance end mill designed for machining corners and edges. It features a corner radius of 0.031" and is made of solid carbide with TiAlN coating for wear resistance and heat dissipation. This 1/4" shank end mill has 2 flutes and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and high temp alloys. It has a center cutting design allowing plunge cuts and can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. The end mill has a reach of 1" and overall length of 2-1/2".