Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502751790
Encompass_Preferred
The Grainger 14T032 is a Dayton 1/3 HP 115/230V Dual Voltage ODP Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a dual voltage capability, open drip proof enclosure, and features sleeve bearings. It can be used for a variety of light industrial, commercial, or residential applications requiring 1/3 HP. The motor dimensions are length 7-1/2", width 5-3/4", height 6-5/8" and it weighs 15 pounds. This versatile 1/3 HP dual voltage ODP motor from Dayton would be a good choice for many light duty drive applications.
Catalog No. 50-275-1790 Supplier Grainger Supplier No. 14T032
The Grainger 14T032 is a Dayton 1/3 HP 115/230V Dual Voltage ODP Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a dual voltage capability, open drip proof enclosure, and features sleeve bearings. It can be used for a variety of light industrial, commercial, or residential applications requiring 1/3 HP. The motor dimensions are length 7-1/2", width 5-3/4", height 6-5/8" and it weighs 15 pounds. This versatile 1/3 HP dual voltage ODP motor from Dayton would be a good choice for many light duty drive applications.