Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502748846
Encompass_Preferred
The Corner Radius End Mill from Grainger is a 4-flute end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/2 inch shank diameter and an overall length of 3 inches. The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill is made in the USA by Niagara Cutter and is sold individually.
Catalog No. 50-274-8846 Supplier Grainger Supplier No. 14N074
The Corner Radius End Mill from Grainger is a 4-flute end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/2 inch shank diameter and an overall length of 3 inches. The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill is made in the USA by Niagara Cutter and is sold individually.