Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502751991
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a 4-flute carbide end mill designed for general purpose milling in a variety of materials. It features a corner radius of 0.031" and is available in diameters ranging from 1/8" to 1/2". The end mill has a TiAIN coating for increased tool life and lubricity. It can be run at speeds up to 25,000 RPM with feed rates from 0.001-0.010 IPR depending on the material being machined. Overall, this is a versatile end mill well-suited for light milling applications where a small corner radius is desired.
Catalog No. 50-275-1991 Supplier Grainger Supplier No. 14T033
The Micro 100 Corner Radius End Mill is a 4-flute carbide end mill designed for general purpose milling in a variety of materials. It features a corner radius of 0.031" and is available in diameters ranging from 1/8" to 1/2". The end mill has a TiAIN coating for increased tool life and lubricity. It can be run at speeds up to 25,000 RPM with feed rates from 0.001-0.010 IPR depending on the material being machined. Overall, this is a versatile end mill well-suited for light milling applications where a small corner radius is desired.