Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502751992
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a high-performance end mill designed for machining corners and edges. It features a corner radius geometry that produces a smooth edge finish and eliminates stress concentrations. This 1/4" diameter, 2-flute end mill has a corner radius of 0.031" and is made of solid carbide for wear resistance and long tool life. With a TiAlN coating for heat resistance, this end mill can machine at high speeds in most steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. It has a center cutting design for plunging capability. The Micro 100 Corner Radius End Mill is well-suited for complex contouring and finishing operations.
Catalog No. 50-275-1992 Supplier Grainger Supplier No. 14T034
The Micro 100 Corner Radius End Mill is a high-performance end mill designed for machining corners and edges. It features a corner radius geometry that produces a smooth edge finish and eliminates stress concentrations. This 1/4" diameter, 2-flute end mill has a corner radius of 0.031" and is made of solid carbide for wear resistance and long tool life. With a TiAlN coating for heat resistance, this end mill can machine at high speeds in most steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. It has a center cutting design for plunging capability. The Micro 100 Corner Radius End Mill is well-suited for complex contouring and finishing operations.