Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502751278
Encompass_Preferred
The Micro 100 Corner Radius End Mill AlTiN 14T026 is a 14mm diameter, 2 flute corner radius end mill made of micrograin carbide with an AlTiN coating. It has a corner radius of 0.020" and an overall length of 2-1/2". The end mill has a TiAlN coating that provides heat and wear resistance. It can machine steels, stainless steels, cast iron, aluminum and other non-ferrous materials. The end mill is designed for high performance milling in a wide range of materials.
Catalog No. 50-275-1278 Supplier Grainger Supplier No. 14T026
The Micro 100 Corner Radius End Mill AlTiN 14T026 is a 14mm diameter, 2 flute corner radius end mill made of micrograin carbide with an AlTiN coating. It has a corner radius of 0.020" and an overall length of 2-1/2". The end mill has a TiAlN coating that provides heat and wear resistance. It can machine steels, stainless steels, cast iron, aluminum and other non-ferrous materials. The end mill is designed for high performance milling in a wide range of materials.