Learn More
Grainger Cor Rad End Mill 1/4 Carb 0.0200 rad
Catalog No. 502748674
Encompass_Preferred
The Micro 100 Corner Radius End Mill AlTiN 14N006 is a high performance end mill made by Micro 100. It has a 0.125" corner radius and is made of micrograin carbide with an AlTiN coating for wear resistance and heat dissipation. This 14N006 end mill has 2 flutes and can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. It's well suited for machining hard materials like tool steels, titanium alloys, nickel alloys, and other exotics. The end mill has a 1/4" shank diameter and is 3" overall length for rigidity and tool life. It can machine a corner radius as small as .015" on the workpiece.
Catalog No. 50-274-8674 Supplier Grainger Supplier No. 14N006
The Micro 100 Corner Radius End Mill AlTiN 14N006 is a high performance end mill made by Micro 100. It has a 0.125" corner radius and is made of micrograin carbide with an AlTiN coating for wear resistance and heat dissipation. This 14N006 end mill has 2 flutes and can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. It's well suited for machining hard materials like tool steels, titanium alloys, nickel alloys, and other exotics. The end mill has a 1/4" shank diameter and is 3" overall length for rigidity and tool life. It can machine a corner radius as small as .015" on the workpiece.