Learn More
Grainger Cor Rad End Mill 1.50mm Carb 0.15mm rad
Catalog No. 502751524
Encompass_Preferred
The Corner Radius End Mill from Grainger is a 4-flute end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and an overall length of 2 inches. The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in aluminum, brass, bronze, copper, plastics, and other non-ferrous materials. This end mill can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. It is made in the USA by Niagara Cutter.
Catalog No. 50-275-1524 Supplier Grainger Supplier No. 14K434
The Corner Radius End Mill from Grainger is a 4-flute end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and an overall length of 2 inches. The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in aluminum, brass, bronze, copper, plastics, and other non-ferrous materials. This end mill can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. It is made in the USA by Niagara Cutter.