Learn More
Grainger Cor Rad End Mill 1/8 Carb 0.0100 rad
Catalog No. 502750062
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch cutting diameter. The overall length is 2 inches. It has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. This end mill is made of solid carbide and can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. It features a TiAlN coating for increased tool life and heat resistance. The corner radius design allows for strong cutting edges and smooth wall blends.
Catalog No. 50-275-0062 Supplier Grainger Supplier No. 14R995
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch cutting diameter. The overall length is 2 inches. It has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. This end mill is made of solid carbide and can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. It features a TiAlN coating for increased tool life and heat resistance. The corner radius design allows for strong cutting edges and smooth wall blends.