Learn More
Grainger Cor Rad End Mill 1/8 Carb 0.0100 rad
Catalog No. 502749909
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.001 to 0.004 IPR. It is designed for general purpose milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. The end mill features a TiAlN coating for increased tool life and heat resistance. It produces a smooth corner radius on parts.
Catalog No. 50-274-9909 Supplier Grainger Supplier No. 14R991
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.001 to 0.004 IPR. It is designed for general purpose milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. The end mill features a TiAlN coating for increased tool life and heat resistance. It produces a smooth corner radius on parts.