Learn More
Grainger Cor Rad End Mill 1/8 Carb 0.0100 rad
Catalog No. 502749905
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM. It is designed for general purpose milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. The end mill features TiAlN coating for increased tool life and lubricity. It produces a smooth corner radius on parts.
Catalog No. 50-274-9905 Supplier Grainger Supplier No. 14R990
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM. It is designed for general purpose milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. The end mill features TiAlN coating for increased tool life and lubricity. It produces a smooth corner radius on parts.