Learn More
Grainger Cor Rad End Mill 1/8 Carb 0.0200 rad
Catalog No. 502749961
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.002 to 0.012 inches per tooth. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum and other non-ferrous metals. The end mill features a TiAlN coating for increased tool life and heat resistance. It produces a smooth corner radius on parts.
Catalog No. 50-274-9961 Supplier Grainger Supplier No. 14R993
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM with feed rates from 0.002 to 0.012 inches per tooth. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum and other non-ferrous metals. The end mill features a TiAlN coating for increased tool life and heat resistance. It produces a smooth corner radius on parts.