Learn More
Grainger Cor Rad End Mill 1/8 Carb 0.0200 rad
Catalog No. 502749962
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has a TiAlN coating and 4 flutes. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The end mill provides good chip control and tool life. It can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. The corner radius design allows for stronger cutting edges and smoother wall finishes.
Catalog No. 50-274-9962 Supplier Grainger Supplier No. 14R996
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a length of 2 inches. The end mill has a TiAlN coating and 4 flutes. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The end mill provides good chip control and tool life. It can be run at speeds up to 21,000 RPM with feed rates up to 80 IPM. The corner radius design allows for stronger cutting edges and smoother wall finishes.