Learn More
Grainger Cor Rad End Mill 12.00mm Carb 0.60mm rad
Catalog No. 502748847
Encompass_Preferred
The Micro 100 Corner Radius End Mill with AlTiN coating is a high performance end mill for machining aluminum, titanium and other non-ferrous metals. It has a 0.125" corner radius and is available in diameters ranging from 1/8" to 1/2". The AlTiN coating provides heat resistance and a smooth surface finish. The end mill features a variable helix flute design to reduce chatter and harmonics. It has a center cutting design allowing plunge cuts into a workpiece. The end mill is made from micrograin carbide for strength and tool life. It is well-suited for high speed machining in aerospace, medical and other industries.
Catalog No. 50-274-8847 Supplier Grainger Supplier No. 14M989
The Micro 100 Corner Radius End Mill with AlTiN coating is a high performance end mill for machining aluminum, titanium and other non-ferrous metals. It has a 0.125" corner radius and is available in diameters ranging from 1/8" to 1/2". The AlTiN coating provides heat resistance and a smooth surface finish. The end mill features a variable helix flute design to reduce chatter and harmonics. It has a center cutting design allowing plunge cuts into a workpiece. The end mill is made from micrograin carbide for strength and tool life. It is well-suited for high speed machining in aerospace, medical and other industries.