Learn More
Grainger Cor Rad End Mill 3.00mm Carb 0.50mm rad
Catalog No. 502752005
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in overall lengths of 1 inch to 3 inches. The end mill has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. It is well-suited for machining corners and edges in steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The end mill features Micro 100's proprietary AM100 coating for increased tool life and performance.
Catalog No. 50-275-2005 Supplier Grainger Supplier No. 14M977
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in overall lengths of 1 inch to 3 inches. The end mill has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. It is well-suited for machining corners and edges in steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The end mill features Micro 100's proprietary AM100 coating for increased tool life and performance.