Learn More
Grainger Cor Rad End Mill 3/16 Carb 0.0100 rad
Catalog No. 502751390
Encompass_Preferred
The Corner Radius End Mill from Niagara Cutter is a 4-flute end mill made of solid carbide with a corner radius of 0.031". It has a 0.375" shank diameter and an overall length of 2.5". The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in steels, stainless steels, cast iron, aluminum and other non-ferrous materials. With its corner radius design, it can mill fillets and chamfers without stress concentration. This is a high quality carbide end mill for efficient machining and clean surface finishes.
Catalog No. 50-275-1390 Supplier Grainger Supplier No. 14T012
The Corner Radius End Mill from Niagara Cutter is a 4-flute end mill made of solid carbide with a corner radius of 0.031". It has a 0.375" shank diameter and an overall length of 2.5". The end mill has a TiAIN coating for heat and wear resistance. It is designed for high speed machining in steels, stainless steels, cast iron, aluminum and other non-ferrous materials. With its corner radius design, it can mill fillets and chamfers without stress concentration. This is a high quality carbide end mill for efficient machining and clean surface finishes.