Learn More
Grainger Cor Rad End Mill 3/16 Carb 0.0100 rad
Supplier:  Grainger 14N054
Encompass_Preferred
The Micro 100 Corner Radius End Mill Carbide is a high performance end mill designed for machining corners and edges. It features a corner radius of 0.031" and a 2-flute design for smooth, efficient cutting. The end mill has a TiAlN coating for heat and wear resistance. It is made of solid carbide for strength and durability. The end mill has a 1/4" shank diameter and is available in multiple lengths from 1/2" to 2-1/2". It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, titanium and more. The end mill provides clean, sharp corners and edges.
Catalog No. 50-274-8845