Learn More
Grainger Cor Rad End Mill 3/16 Carb 0.0200 rad
Catalog No. 502751246
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish, carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2 inches, and 2 flutes. The end mill has a TiAIN coating and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and graphite. It is designed for high speed machining with speeds up to 21,000 RPM and feeds up to 0.004 IPR. The end mill provides good chip control and can produce a fine surface finish.
Catalog No. 50-275-1246 Supplier Grainger Supplier No. 14T009
The Micro 100 Corner Radius End Mill is a bright finish, carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2 inches, and 2 flutes. The end mill has a TiAIN coating and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and graphite. It is designed for high speed machining with speeds up to 21,000 RPM and feeds up to 0.004 IPR. The end mill provides good chip control and can produce a fine surface finish.