Learn More
Grainger Cor Rad End Mill 3/32 Carb 0.0100 rad
Catalog No. 502749520
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch overall length. The end mill has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. It is made of solid carbide for wear resistance when milling steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill is designed for high performance slotting, contouring and profiling applications.
Catalog No. 50-274-9520 Supplier Grainger Supplier No. 14R981
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch overall length. The end mill has 2 flutes and can be run at speeds up to 12,000 RPM with feed rates up to 80 IPM. It is made of solid carbide for wear resistance when milling steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill is designed for high performance slotting, contouring and profiling applications.