Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0100 rad
Catalog No. 502750538
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2 inches, and 2 flutes. The end mill has an AlTiN coating and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper alloys, graphite, and plastics. It is designed for high speed machining with recommended speeds of 400-1200 SFM and feeds of 0.002-0.006 IPR based on material. The end mill provides accurate corner radii and can be used for contouring, slotting, shoulder milling, and profiling applications.
Catalog No. 50-275-0538 Supplier Grainger Supplier No. 14T053
The Micro 100 Corner Radius End Mill is a bright finish end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2 inches, and 2 flutes. The end mill has an AlTiN coating and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper alloys, graphite, and plastics. It is designed for high speed machining with recommended speeds of 400-1200 SFM and feeds of 0.002-0.006 IPR based on material. The end mill provides accurate corner radii and can be used for contouring, slotting, shoulder milling, and profiling applications.