Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0200 rad
Catalog No. 502751877
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish end mill with a corner radius of 0.031 inches. It is made of solid carbide and has 2 flutes. The overall length is 2 inches and it has a shank diameter of 0.25 inches. The end mill has an application range of steels, tool steels, stainless steels, cast irons, aluminum, copper, brass, bronze, plastics, and composites. It can be used for slotting, side milling, contouring, and profiling operations. The end mill provides accurate radii at high feed rates.
Catalog No. 50-275-1877 Supplier Grainger Supplier No. 14T074
The Micro 100 Corner Radius End Mill is a bright finish end mill with a corner radius of 0.031 inches. It is made of solid carbide and has 2 flutes. The overall length is 2 inches and it has a shank diameter of 0.25 inches. The end mill has an application range of steels, tool steels, stainless steels, cast irons, aluminum, copper, brass, bronze, plastics, and composites. It can be used for slotting, side milling, contouring, and profiling operations. The end mill provides accurate radii at high feed rates.