Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0200 rad
Catalog No. 502751880
Encompass_Preferred
The Micro 100 Corner Radius End Mill with AlTiN coating is a 14mm diameter, 2 flute end mill with a corner radius of 0.5mm. It has a TiAlN coating for high temperature resistance and hardness. The end mill has a shank diameter of 3/8" and an overall length of 2-1/2". It is made of solid carbide for wear resistance when milling steel, high temp alloys, titanium and nickel based alloys. The corner radius profile allows for strong cutting edges and smooth wall blends.
Catalog No. 50-275-1880 Supplier Grainger Supplier No. 14T066
The Micro 100 Corner Radius End Mill with AlTiN coating is a 14mm diameter, 2 flute end mill with a corner radius of 0.5mm. It has a TiAlN coating for high temperature resistance and hardness. The end mill has a shank diameter of 3/8" and an overall length of 2-1/2". It is made of solid carbide for wear resistance when milling steel, high temp alloys, titanium and nickel based alloys. The corner radius profile allows for strong cutting edges and smooth wall blends.