Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0200 rad
Catalog No. 502753285
Encompass_Preferred
The Micro 100 Corner Radius End Mill from Grainger is a bright finish end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2-1/2 inches, and 2 flutes. This end mill is center cutting and can mill up to 1/4 inch deep. It's designed for high speed machining in aluminum, brass, bronze, plastic, and other non-ferrous materials. The end mill features Micro 100's proprietary ultra-fine grain carbide substrate for strength, durability and wear resistance. It's made in the USA.
Catalog No. 50-275-3285 Supplier Grainger Supplier No. 14T065
The Micro 100 Corner Radius End Mill from Grainger is a bright finish end mill made of solid carbide with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter, overall length of 2-1/2 inches, and 2 flutes. This end mill is center cutting and can mill up to 1/4 inch deep. It's designed for high speed machining in aluminum, brass, bronze, plastic, and other non-ferrous materials. The end mill features Micro 100's proprietary ultra-fine grain carbide substrate for strength, durability and wear resistance. It's made in the USA.