Learn More
Grainger Cor Rad End Mill 6.00mm Carb 0.50mm rad
Catalog No. 502748672
Encompass_Preferred
The Micro 100 Corner Radius End Mill with AlTiN coating is a high performance end mill for machining aluminum, titanium, and other non-ferrous metals. It has a 0.125" corner radius and is available in multiple diameters from 1/8" to 1/2". The AlTiN coating provides heat resistance and a smooth surface finish. This end mill is made of micrograin carbide for strength and tool life. It has 2 flutes and a 30 degree helix angle. The corner radius design allows milling of edges and contours without stress concentration. This is an excellent choice for high speed machining of non-ferrous aerospace alloys.
Catalog No. 50-274-8672 Supplier Grainger Supplier No. 14N092
The Micro 100 Corner Radius End Mill with AlTiN coating is a high performance end mill for machining aluminum, titanium, and other non-ferrous metals. It has a 0.125" corner radius and is available in multiple diameters from 1/8" to 1/2". The AlTiN coating provides heat resistance and a smooth surface finish. This end mill is made of micrograin carbide for strength and tool life. It has 2 flutes and a 30 degree helix angle. The corner radius design allows milling of edges and contours without stress concentration. This is an excellent choice for high speed machining of non-ferrous aerospace alloys.