Learn More
Anton Paar Cora 100 is a handheld Raman analyzer for point-and-shoot identification of unknown substances. Easily verify hazardous materials, narcotics, explosives, and incoming goods. Intuitive software supports fast, on-site decisions in critical environments
Catalog No. 5000580530
Encompass_Preferred
Technical Specifications Cora 100Laser excitation: 785 nmLaser output power: 300 mW output, >215 mW at sampleSpectral range: 400 cm-1 to 2300 cm-1Spectral resolution: max. 10 cm-1Detector type: Linear CCD arrayDisplay: Transmissive color TFT with LED backlightingSoftware: Embedded LinuxData formats: TextConnectivity: USB 2.0 mass storage deviceBattery: Rechargeable Li-ion, normally 60 scans endurance before recharging1, non-user replaceableAC adapterOptional output 5 VDC/1 A USB for chargingSize: 15.8 cm x 10.1 cm x 2.9 cmWeight: 650 gOperating temperature: -20 °C to +40 °CStorage temperature: -30 °C to +50 °CAccessories: Point-and-shoot adapter, vial holder, 90° angle adapter with mounting tools, laser aperture cap with polystyrene target for system test.Included items with product: Instrument, watertight case, shoulder strap, USB cable, vials
Catalog No. 50-005-80530 Supplier Anton Paar Supplier No. 199171
Please contact your sales representative for a quote or further ordering instructions.
Technical Specifications Cora 100Laser excitation: 785 nmLaser output power: 300 mW output, >215 mW at sampleSpectral range: 400 cm-1 to 2300 cm-1Spectral resolution: max. 10 cm-1Detector type: Linear CCD arrayDisplay: Transmissive color TFT with LED backlightingSoftware: Embedded LinuxData formats: TextConnectivity: USB 2.0 mass storage deviceBattery: Rechargeable Li-ion, normally 60 scans endurance before recharging1, non-user replaceableAC adapterOptional output 5 VDC/1 A USB for chargingSize: 15.8 cm x 10.1 cm x 2.9 cmWeight: 650 gOperating temperature: -20 °C to +40 °CStorage temperature: -30 °C to +50 °CAccessories: Point-and-shoot adapter, vial holder, 90° angle adapter with mounting tools, laser aperture cap with polystyrene target for system test.Included items with product: Instrument, watertight case, shoulder strap, USB cable, vials