Learn More
Anton Paar Cora 5001 fiber raman spectrometer wavelength 1064nm
Catalog No. 5000580449
Encompass_Preferred
The fiber proble need to be selected according to the application, and the wavenght of the Cora 5001 Fiber Raman Spectrometer.
Here is the list of Fiber Probes Available:
235342 RAMAN FIBER PROBE 532 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 532 NM LASER
235343 RAMAN FIBER PROBE 785 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER
235344 RAMAN FIBER PROBE 1064 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 1064 NM LASER
241613 ASC FIBER PROBE 785 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER IN COMBINATION WITH MONOWAVE
243718 HIGH TEMPERATURE PROBE 532 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 532 NM LASER
243719 HIGH TEMPERATURE PROBE 785 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER
243720 HIGH TEMPERATURE PROBE 1064 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 1064 NM LASER
323556 IMMERSION PROBE 785 NM CORA 5001 FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER
Catalog No. 50-005-80449 Supplier Anton Paar Supplier No. 225833
Please contact your sales representative for a quote or further ordering instructions.
The fiber proble need to be selected according to the application, and the wavenght of the Cora 5001 Fiber Raman Spectrometer.
Here is the list of Fiber Probes Available:
235342 RAMAN FIBER PROBE 532 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 532 NM LASER
235343 RAMAN FIBER PROBE 785 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER
235344 RAMAN FIBER PROBE 1064 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 1064 NM LASER
241613 ASC FIBER PROBE 785 nm FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER IN COMBINATION WITH MONOWAVE
243718 HIGH TEMPERATURE PROBE 532 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 532 NM LASER
243719 HIGH TEMPERATURE PROBE 785 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER
243720 HIGH TEMPERATURE PROBE 1064 NM FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 1064 NM LASER
323556 IMMERSION PROBE 785 NM CORA 5001 FOR ALL CORA 5001 FIBER INSTRUMENTS WITH 785 NM LASER