Learn More
Proteintech Group Inc CoraLite488-conjugated Alpha Tubulin Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866031100UL
Encompass_Preferred
The Alpha Tubulin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Alpha Tubulin. This antibody recognizes human, mouse, rat, canine antigen. The Alpha Tubulin antibody has been validated for the following applications: FC (Intra), IF, WB analysis. This antibody is coralite488.
Catalog No. 50-225-7066