Learn More
Proteintech Group Inc CoraLite488-conjugated ARL13B Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48817711100UL
Encompass_Preferred
The ARL13B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human, mouse, rat, canine antigen. The ARL13B antibody has been validated for the following applications: IF analysis. This antibody is coralite488.
Catalog No. 50-225-6846