Learn More
Proteintech Group Inc CoraLite488-conjugated CHCHD2 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866302100UL
Encompass_Preferred
The CHCHD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CHCHD2. This antibody recognizes human, pig, rat antigen. The CHCHD2 antibody has been validated for the following applications: IF analysis. This antibody is coralite488.
Catalog No. 50-225-7151