Learn More
Proteintech Group Inc CoraLite488-conjugated GFAP Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48860190100UL
Encompass_Preferred
The GFAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GFAP. This antibody recognizes human, mouse, rat, pig antigen. The GFAP antibody has been validated for the following applications: IF analysis. This antibody is coralite488.
Catalog No. 50-225-6919