Learn More
Proteintech Group Inc CoraLite488-conjugated HO-1/HMOX1 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866743100UL
Encompass_Preferred
The HO-1/HMOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HO-1/HMOX1. This antibody recognizes human, mouse, rat, pig antigen. The HO-1/HMOX1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite488.
Catalog No. 50-225-7248