Learn More
Proteintech Group Inc CoraLite488-conjugated Lamin B1 Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866095100UL
Encompass_Preferred
The Lamin B1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Lamin B1. This antibody recognizes human, mouse, rat antigen. The Lamin B1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite488.
Catalog No. 50-225-7093