Learn More
Proteintech Group Inc CoraLite488-conjugated NSE Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866150100UL
Encompass_Preferred
The NSE antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human NSE. This antibody recognizes human, mouse, rat, pig antigen. The NSE antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite488.
Catalog No. 50-225-7111